CH6371380243 Chem Impex International, Inc.

Thr-Thr-Thr-OH

  • MDL number MFCD00238189
  • Cas Number : 56395-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/thr-thr-thr-oh/5059?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/thr-thr-thr-oh/5059?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart