CH6371380246 Chem Impex International, Inc.

Isovaleryl-Phe-Nle-Sta-Ala-Sta-OH

  • MDL number MFCD00238289
  • Cas Number : 98092-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Storage temp. Store at =-4
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/isovaleryl-phe-nle-sta-ala-sta-oh/5069?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/isovaleryl-phe-nle-sta-ala-sta-oh/5069?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart