CH6371380251 Chem Impex International, Inc.

Lysinoalanine�l (diastereomeric mixture: LL + LD)

  • MDL number MFCD00058086
  • Cas Number : 4418-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-181
Synonym H-Lys(DL-2-amino-2-carboxyethyl)-OH 2HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/lysinoalanine-font-size-2-face-symbol-middot-font-2hcl-diastereomeric-mixture-ll-ld/5076?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/lysinoalanine-font-size-2-face-symbol-middot-font-2hcl-diastereomeric-mixture-ll-ld/5076?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart