CH6371380256 Chem Impex International, Inc.

N,N-Dimethyl-histidine methyl ester

  • MDL number MFCD01318773
  • Cas Number : 170227-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N,N-Dimethyl-L-His-OMe
Purity 95
Storage temp. Store at 0-8
Physical Form Slightly yellow viscous oil
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-i-n-i-dimethyl-histidine-methyl-ester/5086?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-i-n-i-dimethyl-histidine-methyl-ester/5086?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart