CH6371380257 Chem Impex International, Inc.

N-Acetylindol-3-yl acetate

  • MDL number MFCD00022890
  • Cas Number : 16800-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-84 �C
Synonym 1,3-Diacetoxyindole
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-acetylindol-3-yl-acetate/5090?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-acetylindol-3-yl-acetate/5090?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart