CH6371380262 Chem Impex International, Inc.

N-Methyl-L-threonine hydrochloride

  • MDL number MFCD06795804
  • Cas Number : 2812-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Me-L-Thr-OH HCl
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-threonine-hydrochloride/5107?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-threonine-hydrochloride/5107?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart