CH6371380263 Chem Impex International, Inc.

N-Methyl-L-valine benzyl ester 4-toluenesulfonate salt

  • MDL number MFCD00238324
  • Cas Number : 42492-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-122
Synonym N-Me-L-Val-OBzl TosOH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-valine-benzyl-ester-4-toluenesulfonate-salt/5108?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-valine-benzyl-ester-4-toluenesulfonate-salt/5108?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart