CH6371380267 Chem Impex International, Inc.

N-beta-Aminoethyl-Glycine ethyl ester dihydrochloride

  • MDL number MFCD00672354
  • Cas Number : 24123-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-beta-aminoethyl-glycine-ethyl-ester-dihydrochloride/5125?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-beta-aminoethyl-glycine-ethyl-ester-dihydrochloride/5125?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart