CH6371380268 Chem Impex International, Inc.

N-b-Aminoethyl-Glycine

  • MDL number MFCD00144824
  • Cas Number : 24123-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-150 �C
Synonym N-beta-Aminoethyl-Gly-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-beta-aminoethyl-glycine/5126?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-beta-aminoethyl-glycine/5126?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart