CH6371380270 Chem Impex International, Inc.

Oxythiamine HCl

  • MDL number MFCD00050656
  • Cas Number : 136-16-3
  • Molecular Weight : 338.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8
Physical Form White solid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/oxythiamine·hcl/5131?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/oxythiamine·hcl/5131?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart