CH6371380273 Chem Impex International, Inc.

Phthaloyl-L-Leucine

  • MDL number MFCD00065026
  • Cas Number : 2419-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-125 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form Off-white solid powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/phthaloyl-l-leucine/5137?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/phthaloyl-l-leucine/5137?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart