CH6371380275 Chem Impex International, Inc.

4-Phenylazobenzyloxycarbonyl-Pro-OH

  • MDL number MFCD00238346
  • Cas Number : 194541-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-68 �C
Synonym Pz-L-Pro-OH
Purity 99
Storage temp. Store at 0-8
Physical Form Orange solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/4-phenylazobenzyloxycarbonyl-pro-oh

Safety Information

MSDS http://www.chemimpex.com/4-phenylazobenzyloxycarbonyl-pro-oh
Availability details for
Quantity
  • Availability
  • Add to Cart