CH6371380278 Chem Impex International, Inc.

S-Acetamidomethyl-3-mercaptopropionic acid

  • MDL number MFCD00236748
  • Cas Number : 52574-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym S-Acetamidomethyl-deamino-cysteine
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/s-acetamidomethyl-3-mercaptopropionic-acid/5150?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/s-acetamidomethyl-3-mercaptopropionic-acid/5150?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart