CH6371380281 Chem Impex International, Inc.

b-(4-Methylbenzyl)mercapto-b,b-cyclopenta-methylene-propionic acid

  • MDL number MFCD00080282
  • Cas Number : 87242-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym S-4-Methylbenzyl-pmp
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ß-4-methylbenzyl-mercapto-ß-ß-cyclopenta-methylene-propionic-acid/5155?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ß-4-methylbenzyl-mercapto-ß-ß-cyclopenta-methylene-propionic-acid/5155?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart