CH6371380282 Chem Impex International, Inc.

b-S-(4-Methoxybenzylmercapto)isovaleric acid

  • MDL number MFCD00237153
  • Cas Number : 268219-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-82 �C
Synonym 3-(4-Methoxy-benzylsulfanyl)-3-methyl butyric acid
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/beta-s-4-methoxybenzylmercapto-isovaleric-acid/5156?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/beta-s-4-methoxybenzylmercapto-isovaleric-acid/5156?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart