CH6371380283 Chem Impex International, Inc.

b-S-(4-Methoxybenzylmercapto)-b,b-cyclopentamethylene-propionic acid

  • MDL number MFCD00153490
  • Cas Number : 87242-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [1-(4-Methoxybenzylsulfanyl)cyclohexyl]acetic acid
Purity 99
Storage temp. Store at 0-8 �C
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ß-i-s-i-4-methoxybenzylmercapto-ß-ß-cyclopentamethylene-propionic-acid/5157?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ß-i-s-i-4-methoxybenzylmercapto-ß-ß-cyclopentamethylene-propionic-acid/5157?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart