CH6371380285 Chem Impex International, Inc.

Substance P (9-11)

  • MDL number MFCD00237266
  • Cas Number : 40297-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym H-Gly-Leu-Met-NH2 HCl
Purity 99
Storage temp. Store at =-15
Physical Form Off-white crystalline powder with clumps
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/substance-p-9-11/5159?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/substance-p-9-11/5159?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart