CH6371380286 Chem Impex International, Inc.

Substance P-methyl ester

  • MDL number MFCD00076782
  • Cas Number : 76260-78-1
  • Molecular Weight : 1362.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at =-15°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/substance-p-methyl-ester/5161?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/substance-p-methyl-ester/5161?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart