CH6371380287 Chem Impex International, Inc.

Suc-Ala-Ala-Ala-Ala-Ala-pNA

  • MDL number MFCD00238352
  • Cas Number : 61043-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at =-4
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/suc-ala-ala-ala-ala-ala-pna/5162?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/suc-ala-ala-ala-ala-ala-pna/5162?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart