CH6371380290 Chem Impex International, Inc.

Suc-Ala-Ala-Pro-Arg-pNA

  • MDL number MFCD00238354
  • Cas Number : 131068-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at =-4°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/suc-ala-ala-pro-arg-pna/5165?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/suc-ala-ala-pro-arg-pna/5165?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart