CH6371380291 Chem Impex International, Inc.

Suc-Ala-Glu-Pro-Phe-pNA

  • MDL number MFCD00238367
  • Cas Number : 128802-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at =-4
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/suc-ala-glu-pro-phe-pna/5166?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/suc-ala-glu-pro-phe-pna/5166?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart