CH6371380292 Chem Impex International, Inc.

p-Nitrophenylacetic acid N-hydroxyuccinimide ester

  • MDL number MFCD00047124
  • Cas Number : 68123-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182 - 186
Synonym Succinimidyl-p-nitrophenylacetate
Purity 98
Storage temp. Store at 0-8
Physical Form White to yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-p-i-nitrophenylacetic-acid-n-hydroxyuccinimide-ester/5167?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-p-i-nitrophenylacetic-acid-n-hydroxyuccinimide-ester/5167?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart