CH6371380293 Chem Impex International, Inc.

Succinyl glycine amide

  • MDL number MFCD00238379
  • Cas Number : 200863-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-169° C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/succinyl-glycine-amide/5168?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/succinyl-glycine-amide/5168?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart