CH6371380298 Chem Impex International, Inc.

Na-Tosyl-L-lysine methyl ester hydrochloride

  • MDL number MFCD00038877
  • Cas Number : 5266-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-149� C
Synonym Tos-L-Lys-OMe HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-tosyl-l-lysine-methyl-ester-hydrochloride/5177?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-tosyl-l-lysine-methyl-ester-hydrochloride/5177?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart