CH6371380301 Chem Impex International, Inc.

Trityl-L-phenylalanine diethylammonium salt

  • MDL number MFCD00238394
  • Cas Number : 3226-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Trt-L-Phe-OH DEA
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/trityl-l-phenylalanine-diethylammonium-salt/5183?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/trityl-l-phenylalanine-diethylammonium-salt/5183?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart