CH6371380303 Chem Impex International, Inc.

(Boc-Lys9)-Neurotensin (9-13)-methyl ester

  • MDL number MFCD00237194
  • Cas Number : 89545-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-Lys-Pro-Tyr-Ile-Leu-OMe HCl
Purity 97
Storage temp. Store at 0-8
Physical Form White to slightly pink lyophylisate
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-lys-sup-9-sup-neurotensin-9-13-methyl-ester/5190?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-lys-sup-9-sup-neurotensin-9-13-methyl-ester/5190?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart