CH6371380304 Chem Impex International, Inc.

(R)-3-Isopropylamino-1,2-propanediol

  • MDL number MFCD00237128
  • Cas Number : 97988-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-68 �C
Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-isopropylamino-1-2-propanediol/5194?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-isopropylamino-1-2-propanediol/5194?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart