CH6371380306 Chem Impex International, Inc.

(S)-3-Isopropylamino-1,2-propanediol

  • MDL number MFCD00237129
  • Cas Number : 90742-94-2
  • Molecular Weight : 133.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-61 °C
Purity 95
Storage temp. Store at =-4°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-isopropylamino-1-2-propanediol/5197?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-isopropylamino-1-2-propanediol/5197?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart