CH6371380313 Chem Impex International, Inc.

1-O-Hexadecyl-2-O-methyl-sn-glycerol

  • MDL number MFCD00079347
  • Cas Number : 96960-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-O-Palmityl-2-O-methyl-sn-glycerol
Purity 98
Storage temp. Store at = - 15
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-i-o-i-hexadecyl-2-i-o-i-methyl-i-sn-i-glycerol/5217?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-i-o-i-hexadecyl-2-i-o-i-methyl-i-sn-i-glycerol/5217?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart