CH6371380318 Chem Impex International, Inc.

2-Hydroxy-5-dibenzosuberone

  • MDL number MFCD01318864
  • Cas Number : 17910-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-144
Synonym (2-Hydroxy-10,11-dihydro-5H-dibenzo[a,b]cyclohepten-5-one)
Purity 99
Storage temp. Store at 0-8
Physical Form Greyish powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-hydroxy-5-dibenzosuberone/5235?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-hydroxy-5-dibenzosuberone/5235?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart