CH6371380324 Chem Impex International, Inc.

3-O-Allyl-sn-glycerol

  • MDL number MFCD00236782
  • Cas Number : 55941-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form Colorless to light yellow oil
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-o-allyl-i-sn-i-glycerol/5248?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-o-allyl-i-sn-i-glycerol/5248?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart