CH6371380333 Chem Impex International, Inc.

5-Chloro-2-hydroxy-pyrimidine

  • MDL number MFCD00129723
  • Cas Number : 54326-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Chloropyrimidin-2-ol
Purity 99
Storage temp. Store at 0-8
Physical Form Pale yellow crystals
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-chloro-2-hydroxy-pyrimidine/5272?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-chloro-2-hydroxy-pyrimidine/5272?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart