CH6371380334 Chem Impex International, Inc.

2-Abz-Gly-OH�

  • MDL number MFCD00237464
  • Cas Number : 256657-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-abz-gly-oh-font-size-2-face-symbol-middot-font-hcl/5273?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-abz-gly-oh-font-size-2-face-symbol-middot-font-hcl/5273?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart