CH6371380336 Chem Impex International, Inc.

Anisindione

  • MDL number MFCD00176194
  • Cas Number : 117-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-157
Synonym 2-p-Anisyl-1,3-indandione
Purity 98
Storage temp. Store at 0-8
Physical Form Slightly beige or yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/anisindione/5277?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/anisindione/5277?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart