CH6371380339 Chem Impex International, Inc.

Boc-a-methyl-DL-Valine

  • MDL number MFCD00798615
  • Cas Number : 139938-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-alpha-Me-DL-Val-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-a-methyl-dl-valine/5282?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-a-methyl-dl-valine/5282?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart