CH6371380340 Chem Impex International, Inc.

L-Cystine bis-amide dihydrochloride

  • MDL number MFCD00237450
  • Cas Number : 22671-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 223 �dec.)
Synonym (H-L-Cys-NH2)2 2HCl
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-cystine-bis-amide-dihydrochloride/5286?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-cystine-bis-amide-dihydrochloride/5286?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart