CH6371380343 Chem Impex International, Inc.

L-Cystine bis(tert-butyl ester) dihydrochloride

  • MDL number MFCD00237452
  • Cas Number : 38261-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (H-L-Cys-OtBu)2 2HCl
Purity 97
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-cystine-bis-i-tert-i-butyl-ester-dihydrochloride/5289?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-cystine-bis-i-tert-i-butyl-ester-dihydrochloride/5289?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart