CH6371380346 Chem Impex International, Inc.

1,2-Dilauroyl-sn-glycerol

  • MDL number MFCD00067469

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-alpha,beta-Dilaurin
Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-2-dilauroyl-i-sn-i-glycerol/5298?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-2-dilauroyl-i-sn-i-glycerol/5298?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart