CH6371380353 Chem Impex International, Inc.

1,2-Distearoyl-rac-glycero-3-phosphocholine

  • MDL number MFCD00067532
  • Cas Number : 816-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym DL-beta,gamma-Distearoyl-alpha-lecithin
Purity 98
Storage temp. Store at 0-8
Physical Form White powder with lumps
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-2-distearoyl-i-rac-i-glycero-3-phosphocholine/5310?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-2-distearoyl-i-rac-i-glycero-3-phosphocholine/5310?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart