CH6371380356 Chem Impex International, Inc.

1,2-O-Dihexadecyl-rac-glycerol

  • MDL number MFCD00048476
  • Cas Number : 13071-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-2-i-o-i-dihexadecyl-i-rac-i-glycerol/5315?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-2-i-o-i-dihexadecyl-i-rac-i-glycerol/5315?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart