CH6371380357 Chem Impex International, Inc.

1,2-O-Dihexadecyl-sn-glycerol

  • MDL number MFCD00067472
  • Cas Number : 67337-03-5
  • Molecular Weight : 540.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49-53 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-2-o-dihexadecyl-i-sn-i-glycerol/5317?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-2-o-dihexadecyl-i-sn-i-glycerol/5317?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart