CH6371380358 Chem Impex International, Inc.

1,2-O-Dioctadecyl-rac-glycerol

  • MDL number MFCD00171462
  • Cas Number : 13071-61-9
  • Molecular Weight : 597.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/1-2-o-dioctadecyl-i-rac-i-glycerol/5318?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-2-o-dioctadecyl-i-rac-i-glycerol/5318?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart