CH6371380359 Chem Impex International, Inc.

1,2-O-Dioctadecyl-sn-glycerol

  • MDL number MFCD00236963
  • Cas Number : 82188-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60 - 62 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-2-o-dioctadecyl-i-sn-i-glycerol/5319?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-2-o-dioctadecyl-i-sn-i-glycerol/5319?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart