CH6371380361 Chem Impex International, Inc.

4-Aminopiperidine-4-carboxylic acid 2HCl

  • MDL number MFCD02259522
  • Cas Number : 76508-73-1
  • Molecular Weight : 217.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/4-aminopiperidine-4-carboxylic-acid-font-size-2-face-symbol-middot-font-2hcl/5322?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-aminopiperidine-4-carboxylic-acid-font-size-2-face-symbol-middot-font-2hcl/5322?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart