CH6371380362 Chem Impex International, Inc.

Cinnamoyl-(trans)-L-phenylalanine

  • MDL number MFCD00057835
  • Cas Number : 4950-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-164
Synonym Cinnamoyl-(trans)-L-Phe-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cinnamoyl-i-trans-i-l-phenylalanine/5325?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cinnamoyl-i-trans-i-l-phenylalanine/5325?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart