CH6371380363 Chem Impex International, Inc.

Cyclo (L-Ala-L-ala)

  • MDL number MFCD00190738
  • Cas Number : 5845-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Cyclo(-Ala-Ala)
Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cyclo-l-ala-l-ala/5326?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cyclo-l-ala-l-ala/5326?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart