CH6371380365 Chem Impex International, Inc.

Cyclo(L-Ala-L-His)

  • MDL number MFCD00237604
  • Cas Number : 54300-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at RT
Physical Form Beige powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cyclo-l-ala-l-his/5328?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cyclo-l-ala-l-his/5328?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart