CH6371380367 Chem Impex International, Inc.

Cyclo(-Asp-Gly)

  • MDL number MFCD00237607
  • Cas Number : 52661-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Cyclo(L-Asp-L-Gly)
Storage temp. Store at =-4
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cyclo-asp-gly/5331?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cyclo-asp-gly/5331?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart