CH6371380368 Chem Impex International, Inc.

Cyclo(-D-Ala-D-Ala)

  • MDL number MFCD00236939
  • Cas Number : 23927-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystal or powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cyclo-d-ala-d-ala/5333?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cyclo-d-ala-d-ala/5333?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart