CH6371380369 Chem Impex International, Inc.

Cyclo(Gly-L-Gln)

  • MDL number MFCD00237610
  • Cas Number : 52662-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Cyclo(-Gly-Gln)
Purity 98
Storage temp. Store at 0-8
Physical Form White crystal
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cyclo-gly-l-gln/5338?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cyclo-gly-l-gln/5338?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart